Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Stem Cell Antigen-1 (Sca-1) is a member of the lymphocyte antigen 6 (Ly-6) family. It is expressed on multipotent hematopoietic stem cells (HSC) and is used as a marker of HSC in mice. Sca-1 is used in combination with lineage depletion antibodies to identify murine HSC. Sca-1 can be found in fetal liver, adult bone marrow, and adult peripheral blood and spleen. Sca-1 may be involved in the regulation of B and T cell activation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: locus A/E; Locus AE; Ly-6A.2/Ly-6E.1; Ly-6E; ly6a-e; Ly6A/E; lymphocyte antigen 6 complex; Lymphocyte antigen 6A-2/6E-1; Lymphocyte antigen 6E; SCA-1; Stem cell antigen 1; Stem cell antigen 2; T-cell-activating protein; TAP; Thymic shared antigen 1; TSA-1
Gene Aliases: Ly6; Ly67; Ly6a; Ly6e; Sca-2; Tsa-1
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support