Thermo Scientific Chemicals

4-Methoxyphenol, 98+%, Thermo Scientific Chemicals

Catalog number: A14845.30
250 g, Each
Thermo Scientific Chemicals

4-Methoxyphenol, 98+%, Thermo Scientific Chemicals

Catalog number: A14845.30
250 g, Each
Quantity
Catalog number: A14845.30
also known as A14845-30
Price (USD)
Price: 42.30
Online price: 35.65
Your price:
Quantity
-

Chemical Identifiers

CAS
150-76-5
Form
Crystals or powder or crystalline powder or flakes
Appearance (Color)
White to cream to pale yellow
Identification (FTIR)
Conforms (non-U.S. specification)
Assay (GC)
≥98.0%

Description

4-Methoxyphenol is an active ingredient and used in dermatology. It is employed as a pharmaceutical drug in skin depigmentation. It is used as polymerization inhibitors. For example, in the radical polymerization of acryaltes and styrene monomers. It is also used as an intermediate in the preparationagrochemicals, liquid crystals. It acts as a stabilizer for the formulation of inks, toners and adhesives. It is mainly used as an additive for textile and leather industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxyphenol is an active ingredient and used in dermatology. It is employed as a pharmaceutical drug in skin depigmentation. It is used as polymerization inhibitors. For example, in the radical polymerization of acryaltes and styrene monomers. It is also used as an intermediate in the preparationagrochemicals, liquid crystals. It acts as a stabilizer for the formulation of inks, toners and adhesives. It is mainly used as an additive for textile and leather industries.

Solubility
Soluble in acetone, ethyl acetate, ethanol, ether, benzene and carbon tetrachloride.

Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed. Incompatible with halogens and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text