Fluorene-2-carboxaldehyde, 99%
Fluorene-2-carboxaldehyde, 99%
Fluorene-2-carboxaldehyde, 99%
Thermo Scientific Chemicals

Fluorene-2-carboxaldehyde, 99%

CAS: 30084-90-3 | C14H10O | 194.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A14911.14
also known as A14911-14
Price (TWD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS30084-90-3
IUPAC Name9H-fluorene-2-carbaldehyde
Molecular FormulaC14H10O
InChI KeyMNQGEQSXFDKAPY-UHFFFAOYSA-N
SMILESO=CC1=CC=C2C(CC3=CC=CC=C23)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormPowder
Assay (GC)≥98.5%
Free acid (titration)≤1.0%
Melting Point (clear melt)80.5-86.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only