tert-Butyl bromoacetate, 98%
tert-Butyl bromoacetate, 98%
tert-Butyl bromoacetate, 98%
Thermo Scientific Chemicals

tert-Butyl bromoacetate, 98%

CAS: 5292-43-3 | C6H11BrO2 | 195.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A14917.09
also known as A14917-09
Price (USD)
36.00
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
36.00
Each
Chemical Identifiers
CAS5292-43-3
IUPAC Nametert-butyl 2-bromoacetate
Molecular FormulaC6H11BrO2
InChI KeyBNWCETAHAJSBFG-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Refractive Index1.4430-1.4470 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
tert-Butyl bromoacetate is used as an intermediate in the production of dyes, pharmaceuticals, agrochemicals and in organic compounds. It finds application as an alkylating agent. It plays an important role to prepare (2-oxo-4-vinyl-azetidin-1-yl)-acetic acid tert-butyl ester and in collagenase inhibitors synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl bromoacetate is used as an intermediate in the production of dyes, pharmaceuticals, agrochemicals and in organic compounds. It finds application as an alkylating agent. It plays an important role to prepare (2-oxo-4-vinyl-azetidin-1-yl)-acetic acid tert-butyl ester and in collagenase inhibitors synthesis.

Solubility
Miscible with ethanol, chloroform and ethyl acetate. Immiscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Becherer, T.; Grunewald, C.; Engelschalt, V.; Multhaup, G.; Risse, T.; Haag, R. Polyglycerol based coatings to reduce non-specific protein adsorption in sample vials and on SPR sensors. Anal. Chim. Acta 2015, 867, 47-55.
  2. Kamalov, M.; Harris, P. W.; Hartinger, C. G.; Miskelly, G. M.; Cooper, G. J.; Brimble, M. A. Physicochemical studies on the copper(II) binding by glycated collagen telopeptides. Org. Biomol. Chem. 2015, 13 (10), 3058-3063.