Copper(II) 2,4-pentanedionate, 98%
Copper(II) 2,4-pentanedionate, 98%
Copper(II) 2,4-pentanedionate, 98%
Thermo Scientific Chemicals

Copper(II) 2,4-pentanedionate, 98%

CAS: 13395-16-9 | C10H14CuO4 | 261.76 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
1000 g
Catalog number A14920.18
also known as A14920-18
Price (USD)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS13395-16-9
IUPAC Namecopper(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC10H14CuO4
InChI KeyQYJPSWYYEKYVEJ-FDGPNNRMSA-L
SMILES[Cu++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Blue or blue-grey to purple
FormPowder
Assay (Redox Titration)≥97.5 to ≤102.5%
Copper(II) 2,4-pentanedionate is used as a catalyst for carbene transfer reactions, coupling reactions and Michael addition reactions. It is also used as a catalyst for the polymerization of olefins and transesterification reactions as well as a PVC stabilizer. Further, it is used as a curing agent for epoxy resins, acrylic adhesives, silicone rubbers. In addition to this, it is used as a lubricant additive and paint drier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) 2,4-pentanedionate is used as a catalyst for carbene transfer reactions, coupling reactions and Michael addition reactions. It is also used as a catalyst for the polymerization of olefins and transesterification reactions as well as a PVC stabilizer. Further, it is used as a curing agent for epoxy resins, acrylic adhesives, silicone rubbers. In addition to this, it is used as a lubricant additive and paint drier.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. The most effective of a number of metal chelates studied in enabling the reduction of aromatic nitro-compounds by sodium borohydride: J. Chem. Soc., Perkin 1, 2409 (1979).
  2. Promotes the coupling of organozinc reagents with allylic halides to give , δ-unsaturated esters: Tetrahedron Lett., 24, 2749 (1983).
  3. Catalyst for the carbenoid reaction of diazoacetate esters with silyl enol ethers to give siloxycyclopropanes: Liebigs Ann. Chem., 512 (1984); Org. Synth. Coll., 9, 573 (1998).
  4. In catalytic amounts promotes the selective O-benzylation of primary alcohols with benzyl chloride, in the presence of secondary alcohols or phenols: Tetrahedron Lett., 44, 8483 (2003). For a brief feature on uses of the reagent in synthesis, see: Synlett, 2859 (2005).
  5. Jia, Y.; Su, J.; Chen, Z.; Tan, K.; Chen, Q.; Cao, Z.; Jiang, Y.; Xie, Z.; Zheng, L. Composition-tunable synthesis of Pt-Cu octahedral alloy nanocrystals from PtCu to PtCu3 via underpotential-deposition-like process and their electro-catalytic properties. RSC Adv. 2015, 5 (23), 18153-18158.
  6. Staniuk, M.; Zindel, D.; van Beek, W.; Hirsch, O.; Kränzlin, N.; Niederberger, M.; Koziej, D. Matching the organic and inorganic counterparts during nucleation and growth of copper-based nanoparticles - in situ spectroscopic studies. CrystEngComm 2015, 17, 6962-6971.