pJTI™ R4 CMV-TO MCS pA Vector
pJTI™ R4 CMV-TO MCS pA Vector
The pJTI™ R4 CMV-TO MCS pA vector is designed for the expression of your gene of interest under the controlRead more
Have Questions?
Catalog NumberQuantity
A15004100 μg
Catalog number A15004
Price (USD)
1,682.00
Each
Quantity:
100 μg
Price (USD)
1,682.00
Each
The pJTI™ R4 CMV-TO MCS pA vector is designed for the expression of your gene of interest under the control of the tet-inducible CMV promoter after restriction enzyme cloning and retargeting into the genomic R4 site of a Jump-In™ parental cell line.
This vector can be used for inducible or constitutive expression of your gene of interest, depending on which Jump-In™ parental cell line you use. When this vector is retargeted into a Jump-In™ T-REx™ parental cell line, gene expression is controlled by the tet-operon and can be induced by adding doxycycline to the growth media. When used with a Jump-In™ parental cell line such as the Jump-In™ GripTite™ HEK293 cell line, constitutive expression is achieved after retargeting. The R4 sites in the Jump-In™ parental cell lines result in a high retargeting efficiency, requiring less effort and less time than traditional cell engineering methods. Retargeting of Jump-In™ parental cell lines results in creation of an isogenic pool that is sufficient for cell-based experiments without the need for clonal selection. Alternatively, the high retargeting efficiency allows for easy selection of a positive stable clone for expressing your gene of interest.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Antibiotic Resistance BacterialAmpicillin (AmpR)
Constitutive or Inducible SystemInducible
Delivery TypeTransfection
For Use With (Application)Stable Cell Line Development, Targeted Integration, Regulated Expression
Inducing AgentTetracycline
Product TypeMammalian Expression Vector
Quantity100 μg
Selection Agent (Eukaryotic)Geneticin™ (G-418)
VectorJump-In Vectors
Cloning MethodRestriction Enzyme/MCS
Product LineJump-In
PromoterCMV/TO
Protein TagUntagged
Unit SizeEach
Contents & Storage
pJTI™ R4 CMV-TO MCS pA vector (100 μg), store at -20 °C