This kit contains a rabbit Tb-anti-GST antibody—an equivalent kit containing a goat-sourced antibody is also available.

LanthaScreen® TR-FRET RXR beta Coactivator Assay Kit, rabbit

Catalog number: A15120

 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This kit contains Rabbit Tb-Anti-GST antibody; other kit components are the same as kit PV4385:

The LanthaScreen® TR-FRET Retinoid X Receptor (RXR) beta Coactivator Assay Kit provides a sensitive and robust method for high-throughput screening of potential RXR beta ligands as agonists of coactivator recruitment. The homogeneous mix-and-read assay format results in a ligand EC50 composite value representing the amount of ligand required to bind to receptor, effect a conformational change, and recruit coactivator peptide (Figure 1).

How it works

The LanthaScreen® TR-FRET RXR beta Coactivator Assay Kit includes a terbium (Tb)-labeled anti-GST antibody, a fluorescein-labeled coactivator peptide, and a human RXR beta ligand-binding domain (RXR beta-LBD) that is tagged with glutathione-S-transferase (GST). To assay, the RXR beta-LBD is added to ligand test compounds, followed by addition of a mixture of the fluorescein-coactivator peptide and Tb-anti-GST antibody. After room temperature incubation, the TR-FRET 520:495 emission ratio is calculated and used to determine the EC50 from a dose response curve of the compound

Contents and Storage:

The LanthaScreen® TR-FRET RXR beta Coactivator Assay Kit contains RXR beta-LBD (GST) protein, fluorescein-labeled D22 coactivator peptide, Tb-anti-GST antibody, and buffers. Store components as indicated in the assay protocol (-80°C, -20°C, or +4°C).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Green
Detection Method: Fluorescent
Druggable Target: Nuclear Receptors
For Use With (Equipment): Microplate Reader
Format: 384-well plate
Label or Dye: Tb (Terbium)
Product Line: LanthaScreen®
Product Size: 800 x 20 µL assays
Readout: TR-FRET (Activity)
Technique: TR-FRET
Validated Application: Nuclear Receptor Assay

Contents & storage

Tb-anti-GST Antibody (Rabbit): Store at -20 °C
RXR beta-LBD, GST: Store at -80 °C
Fluoroscein Coactivator Peptide: Store at -20 °C
TR-FRET Coregulatore Buffer A: Store at 4 °C
1M DTT: Store at -20 °C or -80 °C

Documents

Manuals & protocols