3,5-Dichlorophenylhydrazine hydrochloride, 95%
3,5-Dichlorophenylhydrazine hydrochloride, 95%
3,5-Dichlorophenylhydrazine hydrochloride, 95%
Thermo Scientific Chemicals

3,5-Dichlorophenylhydrazine hydrochloride, 95%

CAS: 63352-99-8 | C6H7Cl3N2 | 213.486 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A15127.06
also known as A15127-06
Price (EUR)
94,50
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
94,50
Each
Chemical Identifiers
CAS63352-99-8
IUPAC Namehydrogen (3,5-dichlorophenyl)hydrazine chloride
Molecular FormulaC6H7Cl3N2
InChI KeyBDXZOJVMTJOAPS-UHFFFAOYSA-N
SMILES[H+].[Cl-].NNC1=CC(Cl)=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Chloride)≥94.0 to ≤106.0%
Appearance (Color)White to cream
FormPowder
3,5-Dichlorophenyl)hydrazine Hydrochloride is a building block that has been used for the synthesis of 2H-benzo[b][1,4]oxazin-3(4H)-one derivatives to be used as and anti-leukemic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dichlorophenyl)hydrazine Hydrochloride is a building block that has been used for the synthesis of 2H-benzo[b][1,4]oxazin-3(4H)-one derivatives to be used as and anti-leukemic agent.

Solubility
Soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Peter J. Duggan; Andris J. Liepa; Laura K. O'Dea; and C. Elisabet Tranberg. Chemical Development of MDL 103371: An N-Methyl-d-Aspartate-Type Glycine Receptor Antagonist for the Treatment of Stroke. Org. Proc. Res. Dev., 2000, 4(6), 477-487.
  2. Timothy J.N. Watson et. al. Phenethyl isothiocyanate inhibits growth of human chronic myeloid leukemia K562 cells via reactive oxygen species generation and caspases. Molecular Medicine Reports, 2014, 10(1), 543-549.