Thermo Scientific Chemicals

2-Dimethylaminoethyl chloride hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: A15134.22
100 g, Each
Thermo Scientific Chemicals

2-Dimethylaminoethyl chloride hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: A15134.22
100 g, Each
Quantity

Chemical Identifiers

CAS
4584-46-7
IUPAC Name
hydrogen (2-chloroethyl)dimethylamine chloride
Molecular Formula
C4H11Cl2N
InChI Key
LQLJZSJKRYTKTP-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CN(C)CCCl
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥98.0 to ≤102.0%
Identification (FTIR)
Conforms
Appearance (Color)
White to pale cream to pale yellow
Water Content (Karl Fischer Titration)
≤1%

Description

2-Chloro-N,N-dimethylethylamine is a widely used as a intermediate and starting reagent for organic synthesis. Also acts as Intermediate for the syntheses of bephenium hydroxynaphthoate, diltiazem, mepyramine and phenyltoloxamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-N,N-dimethylethylamine is a widely used as a intermediate and starting reagent for organic synthesis. Also acts as Intermediate for the syntheses of bephenium hydroxynaphthoate, diltiazem, mepyramine and phenyltoloxamine.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text