4-Chlorobenzoic acid, 98+%
4-Chlorobenzoic acid, 98+%
4-Chlorobenzoic acid, 98+%
4-Chlorobenzoic acid, 98+%
Thermo Scientific Chemicals

4-Chlorobenzoic acid, 98+%

CAS: 74-11-3 | C7H5ClO2 | 156.565 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
250 g
1000 g
產品號碼 A15135.30
亦稱為 A15135-30
價格 (TWD)
-
申請報價
Quantity:
250 g
Request bulk or custom format
化學識別
CAS74-11-3
IUPAC Name4-chlorobenzoic acid
Molecular FormulaC7H5ClO2
InChI KeyXRHGYUZYPHTUJZ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(Cl)C=C1
檢視更多
規格Specification Sheet規格表
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Assay (Silylated GC)≥98.0%
Identification (FTIR)Conforms
Melting Point (clear melt)238.0-244.0?C
檢視更多
4-Chlorobenzoic acid is used to determine the diversity of 2-, 3- and 4-chlorobenzoate degraders in two pristine soils with similar physical and chemical characteristics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzoic acid is used to determine the diversity of 2-, 3- and 4-chlorobenzoate degraders in two pristine soils with similar physical and chemical characteristics.

Solubility
Soluble in water(0.08g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Hideko Koshima.; Masaki Nagano.; Toru Asahi. Optical Activity Induced by Helical Arrangements of Tryptamine and 4-Chlorobenzoic Acid in Their Cocrystal. J. Am. Chem. Soc. 2008, 127, (8), 2455-2463.
  2. Fernando Rived.; Martı Roses.; Elisabeth Bosch. Dissociation constants of neutral and charged acids in methyl alcohol. The acid strength resolution. Analytica Chimica Acta. 1998, 374, (2-3), 309-324.