This kit contains a rabbit Tb-anti-GST antibody—an equivalent kit containing a goat-sourced antibody is also available.

LanthaScreen® TR-FRET ROR alpha Coactivator Assay Kit, rabbit

Catalog number: A15146

 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This kit contains Rabbit Tb-Anti-GST antibody; other kit components are the same as kit PV5889:

The LanthaScreen® TR-FRET Retinoid-related Orphan Receptor alpha (ROR alpha) Coactivator Assay Kit provides a sensitive and robust method for high-throughput screening of potential ROR alpha ligands as agonists or inverse agonists of coactivator recruitment. The homogeneous mix-and-read assay uses a terbium (Tb)-labeled anti-GST antibody, a fluorescein-labeled coactivator peptide, and a ROR alpha ligand-binding domain (ROR alpha-LBD) that is tagged with glutathione-S-transferase (GST).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Green
Detection Method: Fluorescent
Druggable Target: Nuclear Receptors
For Use With (Equipment): Microplate Reader
Format: 384-well plate
Label or Dye: Tb (Terbium)
Product Line: LanthaScreen®
Product Size: 800 x 20 µL assays
Readout: TR-FRET (Activity)
Technique: TR-FRET
Validated Application: Nuclear Receptor Assay

Contents & storage

Tb-anti-GST Antibody (Rabbit): Store at -20 °C
ROR alpha-LBD, GST: Store at -80 °C
Fluoroscein Peptide: Store at -20 °C
TR-FRET Coregulator Buffer D: Store at 4 °C
1M DTT: Store at -20 °C or -80 °C

Documents

Manuals & protocols