4-Chlorobenzotrifluoride, 98+%
4-Chlorobenzotrifluoride, 98+%
4-Chlorobenzotrifluoride, 98+%
4-Chlorobenzotrifluoride, 98+%
Thermo Scientific Chemicals

4-Chlorobenzotrifluoride, 98+%

CAS: 98-56-6 | C7H4ClF3 | 180.554 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A15154.0E
also known as A15154-0E
Price (EUR)
259,00
Each
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (EUR)
259,00
Each
Add to cart
Chemical Identifiers
CAS98-56-6
IUPAC Name1-chloro-4-(trifluoromethyl)benzene
Molecular FormulaC7H4ClF3
InChI KeyQULYNCCPRWKEMF-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Assay (GC)≥98.0%
Appearance (Color)Clear colorless
Refractive Index1.4445-1.4485 @ 20?C
FormLiquid
4-Chlorobenzotrifluoride acts as an important intermediate for dyes. It is used as an active component of polyurethane finishes. It is also used in adhesives, pigments, sealant chemicals, paints and coatings. Further, it is used as an environmental friendly solvent for cleaning or degreasing and dielectric fluid. It has a high capacity to dissolve inks, which is used in the printing industry. In addition to this, it is also used as xylene replacement in cleaners, thinners and other aromatic hydrocarbon blends.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzotrifluoride acts as an important intermediate for dyes. It is used as an active component of polyurethane finishes. It is also used in adhesives, pigments, sealant chemicals, paints and coatings. Further, it is used as an environmental friendly solvent for cleaning or degreasing and dielectric fluid. It has a high capacity to dissolve inks, which is used in the printing industry. In addition to this, it is also used as xylene replacement in cleaners, thinners and other aromatic hydrocarbon blends.

Solubility
Miscible with chloroform. Slightly miscible with water.

Notes
Store in a cool place. Light sensitive. Incompatible with sodium dimethyl sulfonate, strong bases and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Liu, H.; Li, X.; Liu, F.; Tan, Y.; Jiang, Y. A simple and novel amide ligand based on quinoline derivative used for palladium-catalyzed Suzuki coupling reaction. J. Organomet. Chem. 2015, 794, 27-32.
  2. Qian, Y. Y.; Lee, M. H.; Yang, W.; Chan, K. S. Aryl carbon-chlorine (Ar-Cl) and aryl carbon-fluorine (Ar-F) bond cleavages by rhodium porphyrins. J. Organomet. Chem. 2015, 791, 82-89.