SensiCell™ DMEM - FAQs
SensiCell™ DMEM - FAQs
View additional product information for SensiCell™ DMEM - FAQs (A1515401)
1 product FAQs found
What alternative do you offer for the discontinued SensiCell DMEM?
The alternative product is Advanced DMEM (Cat. No. 12491-015). Please add 4 mM GlutaMAX Supplement to the medium.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.