Thermo Scientific Chemicals

DL-Serine, 99%

Catalog number: A15184.22
100 g, Each
Thermo Scientific Chemicals

DL-Serine, 99%

Catalog number: A15184.22
100 g, Each
Quantity
Catalog number: A15184.22
also known as A15184-22
Price (USD)
Quantity
-

Chemical Identifiers

CAS
302-84-1
IUPAC Name
2-amino-3-hydroxypropanoic acid
Molecular Formula
C3H7NO3
InChI Key
MTCFGRXMJLQNBG-UHFFFAOYNA-N
SMILES
NC(CO)C(O)=O
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Identification (FTIR)
Conforms

Description

DL-Serine is used in the synthesis of novel tryptoline derivatives as IDO (indoleamine 2,3-Deoxygenase) inhibitors, for potential use in Alzheimer’s treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Serine is used in the synthesis of novel tryptoline derivatives as IDO (indoleamine 2,3-Deoxygenase) inhibitors, for potential use in Alzheimer’s treatment.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text