9-Anthracenemethanol, 97%
9-Anthracenemethanol, 97%
9-Anthracenemethanol, 97%
9-Anthracenemethanol, 97%
Thermo Scientific Chemicals

9-Anthracenemethanol, 97%

CAS: 1468-95-7 | C15H12O | 208.26 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
10 g
50 g
250 g
產品號碼 A15188.18
亦稱為 A15188-18
價格 (TWD)
-
申請報價
Quantity:
50 g
Request bulk or custom format
化學識別
CAS1468-95-7
IUPAC Name(anthracen-9-yl)methanol
Molecular FormulaC15H12O
InChI KeyJCJNNHDZTLRSGN-UHFFFAOYSA-N
SMILESOCC1=C2C=CC=CC2=CC2=CC=CC=C12
檢視更多
規格Specification Sheet規格表
FormCrystals or powder or crystalline powder
Appearance (Color)Yellow to green
Assay (HPLC)≥96.0%
Identification (FTIR)Conforms
Melting Point (clear melt)156.0-167.0?C
An anthracene derivative used in Diels-Alder reactions. Also used in the preparation of 9-anthracenylmethyl-1-piperazinecarboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An anthracene derivative used in Diels-Alder reactions. Also used in the preparation of 9-anthracenylmethyl-1-piperazinecarboxylate.

Solubility
Soluble in hot methanol very faint turbidity.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Roh Y-M, et al. Development of a new approach for total isocyanate determination using the reagent 9-anthracenylmethyl-1-piperazinecarboxylate. Analyst, 2000, 125(9), 1691-96.
  2. Chen-Yu Li; Chen-Yen Tsai; Chia-Her Lin; Bao-Tsan Ko. Synthesis, structural characterization and reactivity of aluminum complexes supported by benzotriazole phenoxide ligands: air-stable alumoxane as an efficient catalyst for ring-opening polymerization of L-lactide. Dalton Transactions (Print Edition), 2011, 40(9), 1880-1887.
  3. For use of the 9-anthrylmethyl group for carboxyl protection, see 9-(Chloromethyl) anthracene, A13579.