9(10H)-Acridone, 99%
9(10H)-Acridone, 99%
9(10H)-Acridone, 99%
Thermo Scientific Chemicals

9(10H)-Acridone, 99%

CAS: 578-95-0 | C13H9NO | 195.221 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A15222.14
also known as A15222-14
Price (EUR)
199,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
199,00
Each
Chemical Identifiers
CAS578-95-0
IUPAC Name9,10-dihydroacridin-9-one
Molecular FormulaC13H9NO
InChI KeyFZEYVTFCMJSGMP-UHFFFAOYSA-N
SMILESO=C1C2=CC=CC=C2NC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to green
FormPowder
Assay (HPLC)≥98.5%
9(10H)-Acridone is used in the preparation of methyl 9,10-dihydro-9-oxoacridine-10-pentanoate, use for dyeing the organic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9(10H)-Acridone is used in the preparation of methyl 9,10-dihydro-9-oxoacridine-10-pentanoate, use for dyeing the organic materials.

Solubility
Insoluble in benzene, chloroform, ether, water and ethanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Ramesh Kumar.; Meena Kumari. Chemistry of Acridone and its analogues: A review. J. Chem. Pharm. Res. 2011, 3, (1), 217-230.
  2. Masaaki Mitsui.; Yasuhiro Ohshima. Structure and Dynamics of 9(10H)-Acridone and Its Hydrated Clusters. I. Electronic Spectroscopy. J. Phys. Chem. A. 2000, 104, (38), 8638-8648.