1,3-Diphenylguanidine, 97%
1,3-Diphenylguanidine, 97%
1,3-Diphenylguanidine, 97%
1,3-Diphenylguanidine, 97%
Thermo Scientific Chemicals

1,3-Diphenylguanidine, 97%

CAS: 102-06-7 | C13H13N3 | 211.268 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 g
250 g
1000 g
產品號碼 A15234.30
亦稱為 A15234-30
價格 (TWD)
-
申請報價
Quantity:
250 g
Request bulk or custom format
化學識別
CAS102-06-7
IUPAC NameN,N''-diphenylguanidine
Molecular FormulaC13H13N3
InChI KeyOWRCNXZUPFZXOS-UHFFFAOYSA-N
SMILESNC(NC1=CC=CC=C1)=NC1=CC=CC=C1
檢視更多
規格Specification Sheet規格表
Melting Point (clear melt)142.0-152.0?C
Appearance (Color)White to pale cream to pale brown
FormPowder
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
1,3-Diphenylguanidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diphenylguanidine is used as pharmaceutical intermediate.

Solubility
Soluble in alcohol, chloroform, hot benzene, hot toluene, and dilute mineral acids

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. P.V.Shah; M.R.Sumler; Y.M.Ioannou; H.L.Fisher; L.L.Hall. Dermal absorption and disposition of 1,3-diphenylguanidine in rats. Journal of Toxicology and Environmental Health. 1985, 15,(5), 623-633.
  2. P.V.Gushchin; M.Haukka; N.A.Bokach; V.Yu.Kukushkin. Platinum(iv)-mediated nucleophilic addition of 1,3-diphenylguanidine to propiononitrile. Russian Chemical Bulletin. 2008, 57,(10), 2125-2131.