Thermo Scientific Chemicals

Cyclopentanol, 99%, Thermo Scientific Chemicals

Catalog number: A15249.0F
2500 mL, Each
Thermo Scientific Chemicals

Cyclopentanol, 99%, Thermo Scientific Chemicals

Catalog number: A15249.0F
2500 mL, Each
Quantity
Catalog number: A15249.0F
also known as A15249-0F
Price (USD)
Price: 352.00
Online price: 299.65
Your price:
Quantity
-

Chemical Identifiers

CAS
96-41-3
IUPAC Name
cyclopentanol
Molecular Formula
C5H10O
InChI Key
XCIXKGXIYUWCLL-UHFFFAOYSA-N
SMILES
OC1CCCC1
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
Refractive Index
1.4515-1.4545 @ 20?C
Form
Liquid
Assay (GC)
≥98.5%

Description

Cyclopentanol is a versatile compound used as a building block, intermediate and solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentanol is a versatile compound used as a building block, intermediate and solvent.

Solubility
Slightly soluble in water. Also soluble in ether, alcohols and acetone.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text