Indole-3-butyric acid, 98%
Indole-3-butyric acid, 98%
Indole-3-butyric acid, 98%
Thermo Scientific Chemicals

Indole-3-butyric acid, 98%

CAS: 133-32-4 | C12H13NO2 | 203.241 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15260.06
also known as A15260-06
Price (USD)
49.50
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
49.50
Each
Chemical Identifiers
CAS133-32-4
IUPAC Name4-(1H-indol-3-yl)butanoic acid
Molecular FormulaC12H13NO2
InChI KeyJTEDVYBZBROSJT-UHFFFAOYSA-N
SMILESOC(=O)CCCC1=CNC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to pale brown to brown or pink
FormPowder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (HPLC)≥97.5%
Identification (FTIR)Conforms
View more
Indole-3-butyric acid is auxin-family plant hormone (phytohormone). IBA is thought to be a precursor of indole-3-acetic acid (IAA) the most abundant and the basic auxin natively occurring and functioning in plants. IAA generates the majority of auxin effects in intact plants, and is the most potent native auxin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole-3-butyric acid is auxin-family plant hormone (phytohormone). IBA is thought to be a precursor of indole-3-acetic acid (IAA) the most abundant and the basic auxin natively occurring and functioning in plants. IAA generates the majority of auxin effects in intact plants, and is the most potent native auxin.

Solubility
Soluble in water(0.25g/L).

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Lucia C. Strader.; Bonnie Bartel. Transport and Metabolism of the Endogenous Auxin Precursor Indole-3-Butyric Acid. Molecular Plant. 2011, 4, (3), 477-486.
  2. Jutta Ludwig-Muller. Indole-3-butyric acid in plant growth and development. Plant Growth Regulation. 2000, 32, (2), 219-230.