Acetato de furfurilo, 97 %, Thermo Scientific Chemicals
Acetato de furfurilo, 97 %, Thermo Scientific Chemicals
Acetato de furfurilo, 97 %, Thermo Scientific Chemicals
Acetato de furfurilo, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetato de furfurilo, 97 %, Thermo Scientific Chemicals

CAS: 623-17-6 | C7H8O3 | 140.138 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
25 g
100 g
Número de catálogo A15285.14
también denominado A15285-14
Precio (USD)
-
Cantidad:
25 g
Pedido a granel o personalizado
Identificadores químicos
CAS623-17-6
IUPAC Name(furan-2-yl)methyl acetate
Molecular FormulaC7H8O3
InChI KeyCKOYRRWBOKMNRG-UHFFFAOYSA-N
SMILESCC(=O)OCC1=CC=CO1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Assay (GC)≥96.0%
Refractive Index1.4605-1.4655 @ 20?C
Appearance (Color)Clear colorless to yellow
FormLiquid
Furfuryl acetate is a flavoring ingredient. It was used in the synthesis of 5-acetoxymethyl-2-vinylfuran and 5-hydroxymethyl-2-vinylfuran via Vilsmeier-Haack and Wittig reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
El acetato de furfurilo es un ingrediente aromatizante. Se empleaba en la síntesis de 5-acetoximetil-2-vinilfurano e 5-hidroximetil-2-vinilfurano por medio de las reacciones de Vilsmeier-Haack y Wittig.

Solubilidad
No miscible en agua.

Notas
Estable en las condiciones de almacenamiento recomendadas. Incompatible con agentes oxidantes.
RUO – Research Use Only

General References:

  1. Alexander Mehner; Ana L.Montero; Ricardo Martinez; Stefan Spange. Synthesis of 5-acetoxymethyl- and 5-hydroxymethyl-2-vinyl-furan. Molecules. 2007, 12,(3), 634-640.
  2. Stephen L.Buchwalter. The polymerization of furfuryl acetate in acetonitrile. Journal of Polymer Science Part A: Polymer Chemistry. 1985, 23,(12), 2897-2911.