Ethylenediaminetetraacetic acid diammonium salt hydrate
Ethylenediaminetetraacetic acid diammonium salt hydrate
Ethylenediaminetetraacetic acid diammonium salt hydrate
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid diammonium salt hydrate

CAS: 20824-56-0 | C10H24N4O9 | 344.321 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A15286.14
または、製品番号A15286-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS194491-30-0
Appearance (Color)White
FormPowder or crystalline solid
Complexometric Titration>96.0 to <104.0% (dry wt. basis)
Water Content (Karl Fischer Titration)<12.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only