Thermo Scientific Chemicals

4-Chlorophenylacetonitrile, 98+%, Thermo Scientific Chemicals

Catalog number: A15295.36
500 g, Each
Thermo Scientific Chemicals

4-Chlorophenylacetonitrile, 98+%, Thermo Scientific Chemicals

Catalog number: A15295.36
500 g, Each
Quantity
Catalog number: A15295.36
also known as A15295-36
Price (USD)
Price: 87.80
Online price: 74.65
Your price:
Quantity
-

Chemical Identifiers

CAS
140-53-4
IUPAC Name
2-(4-chlorophenyl)acetonitrile
Molecular Formula
C8H6ClN
InChI Key
IVYMIRMKXZAHRV-UHFFFAOYSA-N
SMILES
ClC1=CC=C(CC#N)C=C1
Appearance (Color)
Clear colorless or white to pale yellow
Form
Crystalline or fused solid (liquid as melt)
Identification (FTIR)
Conforms
Assay (GC)
≥98.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text