TaqPath™ 1-Step RT-qPCR Master Mix, CG, 1 x 10 mL - FAQs
TaqPath™ 1-Step RT-qPCR Master Mix, CG, 1 x 10 mL - FAQs
View additional product information for TaqPath™ 1-Step RT-qPCR Master Mix, CG - FAQs (A15300, A15299)
1 product FAQs found
What is the recommended RNA input range for TaqPath 1-Step RT-qPCR Master Mix, CG (Cat. Nos. A15299, A15300)?
The figure for TaqPath 1-Step RT-qPCR Master Mix, CG (Cat. Nos. A15299, A15300) below, shows the dynamic range of RNA Triplex Amplification at 0.1 pg–100 ng. While a reaction volume is not specified, we typically use 20 µL reactions.
RNA Triplex Amplification
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.