Coumarin, 98%
Coumarin, 98%
Coumarin, 98%
Thermo Scientific Chemicals

Coumarin, 98%

CAS: 91-64-5 | C9H6O2 | 146.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A15336.0I
also known as A15336-0I
Price (EUR)
507,00
Each
Quantity:
5000 g
Request bulk or custom format
Price (EUR)
507,00
Each
Chemical Identifiers
CAS91-64-5
IUPAC Name2H-chromen-2-one
Molecular FormulaC9H6O2
InChI KeyZYGHJZDHTFUPRJ-UHFFFAOYSA-N
SMILESO=C1OC2=CC=CC=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Melting Point (clear melt)67.0-73.0?C
Coumarin is an anticoagulant found in medicinal plants and used in phytomedicine for the treatment of venous diseases. This compound is found to have anti-tumor activities in several human cells lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Nawal Kishor Mal; Masahiro Fujiwara; Yuko Tanaka. Photocontrolled reversible release of guest molecules from coumarin-modified mesoporous silica. Nature. 2003, 421,350-353.
  2. Guilford JonesII; William R.Jackson; Chol Yoo Choi; William R.Bergmark. Solvent effects on emission yield and lifetime for coumarin laser dyes. Requirements for a rotatory decay mechanism. J. Phys. Chem. 1985, 89,(2), 294-300.