Thermo Scientific Chemicals

Methyl crotonate, 96%, trans-isomer, Thermo Scientific Chemicals

Catalog number: A15363.22
100 g, Each
Thermo Scientific Chemicals

Methyl crotonate, 96%, trans-isomer, Thermo Scientific Chemicals

Catalog number: A15363.22
100 g, Each
Quantity
Catalog number: A15363.22
also known as A15363-22
Price (USD)
Price: 42.00
Online price: 35.65
Your price:
Quantity
-

Chemical Identifiers

CAS
623-43-8
IUPAC Name
methyl (2E)-but-2-enoate
Molecular Formula
C5H8O2
InChI Key
MCVVUJPXSBQTRZ-ONEGZZNKSA-N
SMILES
COC(=O)\C=C\C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥95.0%
Form
Liquid
Refractive Index
1.4225-1.4285 @20?C
Identification (FTIR)
Conforms

Description

Methyl crotonate was used to investigate chemo selectivity in the reaction between methyl crotonate and benzyl amine catalyzed by lipase B from Candida Antarctica using solvent engineering. It was used as starting reagent during the total synthesis of phytotoxins solanapyrones D(1) and E(2).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl crotonate was used to investigate chemo selectivity in the reaction between methyl crotonate and benzyl amine catalyzed by lipase B from Candida Antarctica using solvent engineering. It was used as starting reagent during the total synthesis of phytotoxins solanapyrones D(1) and E(2).

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text