Methyl 4-(trifluoromethyl)benzoate, 98%
Methyl 4-(trifluoromethyl)benzoate, 98%
Methyl 4-(trifluoromethyl)benzoate, 98%
Thermo Scientific Chemicals

Methyl 4-(trifluoromethyl)benzoate, 98%

CAS: 2967-66-0 | C9H7F3O2 | 204.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15400.06
also known as A15400-06
Price (TWD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS2967-66-0
IUPAC Namemethyl 4-(trifluoromethyl)benzoate
Molecular FormulaC9H7F3O2
InChI KeyVAZWXPJOOFSNLB-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4505-1.4545 @ 20?C
FormLiquid
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only