1,8-Octanediol is used in cosmetics, perfumes, ink, essence and in UV coating. It acts as a precursor in the synthesis of polymer and in the manufacture of pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
El 1,8-octandiol se utiliza en cosméticos, perfumes, tinta, esencias y en revestimientos UV. Actúa como precursor en la síntesis de polímeros y en la fabricación de productos farmacéuticos.
Solubilidad
Soluble en agua y metanol.
Notas
Incompatible con agentes oxidantes fuertes, agentes reductores, cloruros ácidos, anhídridos ácidos y cloroformiatos.
RUO – Research Use Only
General References:
- Qian, F.; Luo, Q.; Yang, R.; Zhu, Z.; Chen, H.; Yan, X. The littoral red alga Pyropia haitanensisuses rapid accumulation of floridoside as the desiccation acclimation strategy. J. Appl. Phycol. 2015, 27 (1), 621-632.
- Zeimaran, E.; Pourshahrestani, S.; Pingguan-Murphy, B.; Kadri, N. A.; Rothan, H. A.; Yusof, R.; Towler, M. R.; Djordjevic, I. Fabrication and characterization of poly(octanediol citrate)/gallium-containing bioglass microcomposite scaffolds. J. Mater. Sci. 2015, 50 (5), 2189-2201.