Thermo Scientific Chemicals

N-Acetyl-L-cysteine, 98+%

Catalog number: A15409.14
25 g, Each
Thermo Scientific Chemicals

N-Acetyl-L-cysteine, 98+%

Catalog number: A15409.14
25 g, Each
Quantity
Catalog number: A15409.14
also known as A15409-14
Price (USD)

Chemical Identifiers

CAS
616-91-1
IUPAC Name
2-acetamido-3-sulfanylpropanoic acid
Molecular Formula
C5H9NO3S
InChI Key
PWKSKIMOESPYIA-UHFFFAOYNA-N
SMILES
CC(=O)NC(CS)C(O)=O
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0% (UK sourced material)
Comment
Material sourced in UK or US
Loss on Drying
≤0.5% (US sourced material)
Optical Rotation
+5.5? ±1? (c=2 in Water) or +21? to +27? (c=5, phosphate buffer pH 7)
Appearance (Color)
White

Description

A mucolytic agent for isolation of mycobacteria from sputum.N-Acetyl-L-cysteine is used in the liquefaction and decontamination of sputum during the preparation for recovery of mycobacterium, which is called as Petroff's method. It shows antiviral activity against the influenza A viruses. It is used in the treatment of interstitial lung disease to prevent disease progression. It is an antioxidant, dietary supplement and protects liver. Further, it forms complexes with palladium, thereby it dissolves in water and remove palladium from drugs. Further, it is used in the treatment of paracetamol overdose. In addition to this, it is involved in the treatment of autism, Alzheimer's disease, bipolar disorder, major depressive disorder, obsessive-compulsive disorder, schizophrenia, and specific drug addictions like cocaine and cannabis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • N-Acetyl-L-cysteine (acetylcysteine) is a precursor of the antioxidant glutathione, or GSH
  • N-Acetyl-L-cysteine is cytoprotective and may prevent cancer, neurodegeneration, and other disease

Application

  • N-Acetyl-L-cysteine is useful for cellular redox study because of its anti-inflammatory properties
  • It may also be useful for psychiatric research and the study of neurodegeneration
  • It is suitable for in vivo and in vitro experiments
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text