1,1,4,7,7-Pentamethyldiethylenetriamine, 98%
1,1,4,7,7-Pentamethyldiethylenetriamine, 98%
1,1,4,7,7-Pentamethyldiethylenetriamine, 98%
Thermo Scientific Chemicals

1,1,4,7,7-Pentamethyldiethylenetriamine, 98%

CAS: 3030-47-5 | C9H23N3 | 173.304 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A15433.36
also known as A15433-36
Price (USD)
469.65
Online Exclusive
522.00
Save 52.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
469.65
Online Exclusive
522.00
Save 52.35 (10%)
Each
Chemical Identifiers
CAS3030-47-5
IUPAC Name(2-{[2-(dimethylamino)ethyl](methyl)amino}ethyl)dimethylamine
Molecular FormulaC9H23N3
InChI KeyUKODFQOELJFMII-UHFFFAOYSA-N
SMILESCN(C)CCN(C)CCN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4400-1.4450 @ 20?C
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%
FormLiquid
Used as ligand in NMR study of complexation/aggregation with neopentyllithium1 and in the study of ligand effects on the selectivity of CO2 incorporation into α,ω-diynes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as ligand in NMR study of complexation/aggregation with neopentyllithium1 and in the study of ligand effects on the selectivity of CO2 incorporation into α,ω-diynes

Solubility
Miscible in water.

Notes
Air & Moisture Sensitive. Store under inert gas. Incompatible with strong oxidizing agents, strong acids.
RUO – Research Use Only

General References:

  1. L. Banci; A. Bencini; D. Gatteschi. Anisotropic exchange in transition-metal dinuclear complexes. 3. Bis(.mu.-(1,3-azido)bis(1,1,4,7,7-pentamethyldiethylenetriamine)dicopper(II) bis(tetraphenylborate).Inorg. Chem.1984, 23 (14), 2138-2141.
  2. Hou Chen; Delong Liu; Yuting Song; Rongjun Qu and Chunhua Wang. ARGET ATRP of acrylonitrile with ionic liquid as reaction media and 1,1,4,7,7-pentamethyldiethylenetriamine as both ligand and reducing agent in the presence of air.Polymers for Advanced Technologies.2011, 22 1513-1517.