Chromone-3-carboxaldehyde, 97%
Chromone-3-carboxaldehyde, 97%
Chromone-3-carboxaldehyde, 97%
Thermo Scientific Chemicals

Chromone-3-carboxaldehyde, 97%

CAS: 17422-74-1 | C10H6O3 | 174.155 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15438.06
also known as A15438-06
Price (EUR)
34,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
34,00
Each
Chemical Identifiers
CAS17422-74-1
IUPAC Name4-oxo-4H-chromene-3-carbaldehyde
Molecular FormulaC10H6O3
InChI KeyFSMYWBQIMDSGQP-UHFFFAOYSA-N
SMILESO=CC1=COC2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
FormPowder
Appearance (Color)Cream to yellow to pale orange
Melting Point (clear melt)147.0-156.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only