m-Toluenesulfonyl chloride, 98%
m-Toluenesulfonyl chloride, 98%
m-Toluenesulfonyl chloride, 98%
m-Toluenesulfonyl chloride, 98%
Thermo Scientific Chemicals

m-Toluenesulfonyl chloride, 98%

CAS: 1899-93-0 | C7H7ClO2S | 190.641 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
25 g
100 g
Catalog number A15458.22
also known as A15458-22
Price (EUR)
695,00
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
695,00
Each
Add to cart
Chemical Identifiers
CAS1899-93-0
IUPAC Name3-methylbenzene-1-sulfonyl chloride
Molecular FormulaC7H7ClO2S
InChI KeyKFPMLWUKHQMEBU-UHFFFAOYSA-N
SMILESCC1=CC(=CC=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5465-1.5515 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
FormLiquid
Reactant involved in they synthesis of biologically active molecules including small molecule ligands for the Stat3 SH2 domain, aryldisulfonamides with antibacterial activity, quinazoline analogs for the treatment of Gaucher disease, 17β-HSD2 inhibitors for the treatment of osteoporosis, isoindoline-5-propenehydroxamates for use as histone deactylase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents.
RUO – Research Use Only

General References:

  1. Wen-Hsing Lin; Shu-Yi Hsieh; Shih-Chieh Yen; Chiung-Tong Chen; Teng-Kuang Yeh; Tsu Hsu; Cheng-Tai Lu; Ching-Ping Chen; Chun-Wha Chen; Ling-Hui Chou; Yu-Lin Huang; An-Huei Cheng; Yun-I Chang; Ya-Ju Tseng; Kuei-Rong Yen; Yu-Sheng Chao; John T.-A. Hsu; Weir-Torn Jiaang. Discovery and evaluation of 3-phenyl-1H-5-pyrazolylamine-based derivatives as potent, selective and efficacious inhibitors of FMS-like tyrosine kinase-3 (FLT3). Bioorganic & Medicinal Chemistry. 2011, 19 (14), 4173-4182.
  2. Brent D G Page; Steven Fletcher; Peibin Yue; Zhihua Li; Xiaolei Zhang; Sumaiya Sharmeen; Alessandro Datti; Jeffrey L Wrana; Suzanne Trudel; Aaron D Schimmer; James Turkson; Patrick T Gunning. Identification of a non-phosphorylated, cell permeable, small molecule ligand for the Stat3 SH2 domain. Bioorganic & Medicinal Chemistry Letters. 2011, 21 (18), 5605-5609.