Thermo Scientific Chemicals

Di-n-heptyl phthalate, 98+%, Thermo Scientific Chemicals

Catalog number: A15466.18
50 g, Each
Thermo Scientific Chemicals

Di-n-heptyl phthalate, 98+%, Thermo Scientific Chemicals

Catalog number: A15466.18
50 g, Each
Quantity
Catalog number: A15466.18
also known as A15466-18
Price (USD)
Price: 244.00
Online price: 210.65
Your price:
Quantity
-

Chemical Identifiers

CAS
3648-21-3
Refractive Index
1.4835-1.4875 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.0%
Form
Liquid

Description

Di-n-heptyl Phthalate is a Phthalate derivative and an organic extract contaminant found in drinking water, which was shown to activate Nrf-2-Mediated Antioxidant response in human cell line.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-heptyl Phthalate is a Phthalate derivative and an organic extract contaminant found in drinking water, which was shown to activate Nrf-2-Mediated Antioxidant response in human cell line.

Solubility
Sparingly soluble in water.

Notes
Incompatible with Strong acids, strong bases,.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text