Thermo Scientific Chemicals

4-Fluorobenzylamine, 98+%, Thermo Scientific Chemicals

Catalog number: A15486.18
50 g, Each
Thermo Scientific Chemicals

4-Fluorobenzylamine, 98+%, Thermo Scientific Chemicals

Catalog number: A15486.18
50 g, Each
Quantity
Catalog number: A15486.18
also known as A15486-18
Price (USD)
Price: 112.00
Online price: 95.65
Your price:
Quantity
-

Chemical Identifiers

CAS
140-75-0
IUPAC Name
1-(4-fluorophenyl)methanamine
Molecular Formula
C7H8FN
InChI Key
IIFVWLUQBAIPMJ-UHFFFAOYSA-N
SMILES
NCC1=CC=C(F)C=C1
Appearance (Color)
Clear, colourless to pale yellow
Assay (GC)
> 98.0%
Form
liquid
Refractive Index
1.5115 - 1.5155 @20?C

Description

4-Fluorobenzylamine is used as a potassium channel blocking agent. It is used in the synthesis of new tris-iron(III) chelates of 3-hydroxy-4-pyridinone ligands. It reacts with the α- and γ-carboxyl groups of folic acid to yield 18F-labeled folate2. It is an important building block for the synthesis of 18F-labeled compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorobenzylamine is used as a potassium channel blocking agent. It is used in the synthesis of new tris-iron(III) chelates of 3-hydroxy-4-pyridinone ligands. It reacts with the α- and γ-carboxyl groups of folic acid to yield 18F-labeled folate2. It is an important building block for the synthesis of 18F-labeled compounds.

Solubility
Soluble in water.

Notes
Air sensitive. Store under inert gas. Incompatible with oxidizing agent, air, carbon dioxide.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text