Ethyl acrylate, 99%, stab. with ca 20ppm 4-methoxyphenol
Ethyl acrylate, 99%, stab. with ca 20ppm 4-methoxyphenol
Ethyl acrylate, 99%, stab. with ca 20ppm 4-methoxyphenol
Thermo Scientific Chemicals

Ethyl acrylate, 99%, stab. with ca 20ppm 4-methoxyphenol

CAS: 140-88-5 | C5H8O2 | 100.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2 L
Catalog number A15493.AE
also known as A15493-AE
Price (EUR)
32,40
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
32,40
Each
Chemical Identifiers
CAS140-88-5
IUPAC Nameethyl prop-2-enoate
Molecular FormulaC5H8O2
InChI KeyJIGUQPWFLRLWPJ-UHFFFAOYSA-N
SMILESCCOC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Stabilizer4-Methoxyphenol (MEHQ) ca.20ppm
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.5%
Identification (FTIR)Conforms
View more
Ethyl acrylate is used in the syntheses of various pharmaceutical intermediates, textiles and non-woven fibers. It is widely used in the production of polymers including resins, plastics, rubber and denture material. It is actively involved in the preparation of dimethylaminoethylacrylate (DMAEA) which is used as a monomer to make flocculants for water treatment. It plays a vital role in the manufacture of water-based latex paints and adhesives, textile and paper coatings and leather finish resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl acrylate is used in the syntheses of various pharmaceutical intermediates, textiles and non-woven fibers. It is widely used in the production of polymers including resins, plastics, rubber and denture material. It is actively involved in the preparation of dimethylaminoethylacrylate (DMAEA) which is used as a monomer to make flocculants for water treatment. It plays a vital role in the manufacture of water-based latex paints and adhesives, textile and paper coatings and leather finish resins.

Solubility
Miscible with ethanol, ethyl ether and chloroform. Silghtly miscible with water and dimethyl sulfoxide.

Notes
Store in cool place. Incompatible with strong oxidizing agents, strong acids, amines, peroxides, chlorosulfonic acid, moisture and strong alkalies.
RUO – Research Use Only

General References:

  1. Sun, Y.; Zhang, Q.; Hu, J.; Chen, J.; Wang, W. Theoretical study for OH radical-initiated atmospheric oxidation of ethyl acrylate. Chemosphere 2015, 119, 626-633.
  2. del Real, A.; Wallander, D.; Maciel, A.; Cedillo, G.; Loza, H. Graft copolymerization of ethyl acrylate onto tamarind kernel powder, and evaluation of its biodegradability. Carbohydr. Polym. 2015, 117, 11-18.