Aminoacetaldehyde dimethyl acetal, 99%
Aminoacetaldehyde dimethyl acetal, 99%
Aminoacetaldehyde dimethyl acetal, 99%
Thermo Scientific Chemicals

Aminoacetaldehyde dimethyl acetal, 99%

CAS: 22483-09-6 | C4H11NO2 | 105.137 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A15498.14
also known as A15498-14
Price (EUR)
43,50
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
43,50
Each
Chemical Identifiers
CAS22483-09-6
IUPAC Name2,2-dimethoxyethan-1-amine
Molecular FormulaC4H11NO2
InChI KeyQKWWDTYDYOFRJL-UHFFFAOYSA-N
SMILESCOC(CN)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.5%
Refractive Index1.4155-1.4180 @ 20°C
FormLiquid
Amino acetaldehyde dimethyl was used in preparation of chitosan- dendrimer hybrids having various functional groups such as carboxyl, ester and poly(ethylene glycol). It was used in an synthesis of a bicyclic proline analog from L-ascorbic acid and in 3-component reaction catalyzed by MgClO4 leading to α-aminophosphonates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amino acetaldehyde dimethyl was used in preparation of chitosan- dendrimer hybrids having various functional groups such as carboxyl, ester and poly(ethylene glycol). It was used in an synthesis of a bicyclic proline analog from L-ascorbic acid and in 3-component reaction catalyzed by MgClO4 leading to α-aminophosphonates.

Solubility
Miscible in water. Soluble in chloroform, methanol.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
RUO – Research Use Only

General References:

  1. R W Storms; A P Trujillo; J B Springer; L Shah; O M Colvin; S M Ludeman; C Smith. Isolation of primitive human hematopoietic progenitors on the basis of aldehyde dehydrogenase activity. PNAS. 1999, 96 (16), 9118-9123.
  2. Hitoshi Sashiwa; Hirufumi Yajima; Sei-ichi Aiba. Synthesis of a chitosan-dendrimer hybrid and its biodegradation. Biomacromolecules. 2003, 4 (5), 1244-1249.