4-Bromothioanisole, 98+%
4-Bromothioanisole, 98+%
4-Bromothioanisole, 98+%
Thermo Scientific Chemicals

4-Bromothioanisole, 98+%

CAS: 104-95-0 | C7H7BrS | 203.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A15504.36
also known as A15504-36
Price (USD)
992.65
Online Exclusive
1,103.00
Save 110.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
992.65
Online Exclusive
1,103.00
Save 110.35 (10%)
Each
Chemical Identifiers
CAS104-95-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale yellow
FormCrystals or powder or crystalline powder or fused or granular solid
Assay (GC)≥98.0%
Water Content (Karl Fischer Titration)<1.0%
Melting Point (clear melt)37-40?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only