Indium(III) phosphide, polycrystalline lump, 99.99% (metals basis)
Indium(III) phosphide, polycrystalline lump, 99.99% (metals basis)
Indium(III) phosphide, polycrystalline lump, 99.99% (metals basis)
Thermo Scientific Chemicals

Indium(III) phosphide, polycrystalline lump, 99.99% (metals basis)

CAS: 22398-80-7 | InP | 145.792 g/mol
製品番号(カタログ番号) A15509.03
または、製品番号A15509-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS22398-80-7
IUPAC Nameindiganylidynephosphane
Molecular FormulaInP
InChI KeyGPXJNWSHGFTCBW-UHFFFAOYSA-N
SMILESP#[In]
さらに表示
Appearance (Color)Grey to black
FormCrystalline solid
Total Metal Impurities< 100pm
InP is used in high-power and high-frequency electronics because of its superior electron velocity. It also has a direct bandgap, making it useful for optoelectronics devices like laser diodes. It is also used as a substrate for epitaxial indium gallium arsenide based opto-electronic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
InP is used in high-power and high-frequency electronics because of its superior electron velocity. It also has a direct bandgap, making it useful for optoelectronics devices like laser diodes. It is also used as a substrate for epitaxial indium gallium arsenide based opto-electronic devices.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
RUO – Research Use Only

General References:

  1. Roman Shchepin; Dhammika H M L P Navarathna; Raluca Dumitru; Shane Lippold; Kenneth W Nickerson; Patrick H Dussault. InP/ZnS as a safer alternative to CdSe/ZnS core/shell quantum dots: in vitro and in vivo toxicity assessment. Nanoscale. 2013, 5 (1), 307-317.
  2. Xuyong Yang; Dewei Zhao; Kheng Swee Leck; Swee Tiam Tan; Yu Xin Tang; Junliang Zhao; Hilmi Volkan Demir; Xiao Wei Sun. Full visible range covering InP/ZnS nanocrystals with high photometric performance and their application to white quantum dot light-emitting diodes. Biomacromolecules. 2012, 24 (30), 4180-4185.