5-?Bromopentyl Acetate is used to model the toxicity of haloesters on marine bacterium such as Vibrio fischeri. It is used to produce 5-Bromo-pentan-1-ol, a raw material for chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Bromopentyl Acetate is used to model the toxicity of haloesters on marine bacterium such as Vibrio fischeri. It is used to produce 5-Bromo-pentan-1-ol, a raw material for chemical synthesis.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
RUO – Research Use Only