Thermo Scientific Chemicals

Methyl 4-bromocrotonate, tech. 85%, Thermo Scientific Chemicals

Catalog number: A15582.18
50 g, Each
Thermo Scientific Chemicals

Methyl 4-bromocrotonate, tech. 85%, Thermo Scientific Chemicals

Catalog number: A15582.18
50 g, Each
Quantity
Catalog number: A15582.18
also known as A15582-18
Price (USD)
Price: 162.00
Online price: 139.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1117-71-1
IUPAC Name
methyl (2E)-4-bromobut-2-enoate
Molecular Formula
C5H7BrO2
InChI Key
RWIKCBHOVNDESJ-NSCUHMNNSA-N
SMILES
COC(=O)\C=C\CBr
Form
Liquid
Assay (GC)
>83.0%
Appearance (Color)
Clear colorless to yellow
Refractive Index
1.4970-1.5050 @ 20?C

Description

It is a useful synthetic intermediate. It was used in the synthesis of irreversible inhibitors of EGFR and HER-2 tyrosine kinases with enhanced antitumor activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful synthetic intermediate. It was used in the synthesis of irreversible inhibitors of EGFR and HER-2 tyrosine kinases with enhanced antitumor activities.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text