Thermo Scientific Chemicals

1-Bromooctadecane, 97%, Thermo Scientific Chemicals

100g, Each
Thermo Scientific Chemicals

1-Bromooctadecane, 97%, Thermo Scientific Chemicals

100g, Each
Quantity
Catalog number: A15588.22
also known as A15588-22
Price (USD)
Price: 27.90
Your Price:
Quantity
-

Chemical Identifiers

CAS
112-89-0
Assay (GC)
≥96.0%
Appearance (Color)
White to pale cream
Form
Fused solid
Melting Point (clear melt)
25.0-34.0?C

Description

1-Bromooctadecane is used in the preparation of shortened single-walled carbon nanotubes (s-SWCNTs). It is utilized to prepare octadecane in the presence of sodium borohydride as a catalyst. It is involved as a raw material for the preparation of dimethyldistearylammonium bromide, which is a bentonite modifier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromooctadecane is used in the preparation of shortened single-walled carbon nanotubes (s-SWCNTs). It is utilized to prepare octadecane in the presence of sodium borohydride as a catalyst. It is involved as a raw material for the preparation of dimethyldistearylammonium bromide, which is a bentonite modifier.

Solubility
Soluble in acetone and most common organic solvents. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only