Thermo Scientific Chemicals

4-n-Nonylphenol, 98+%, Thermo Scientific Chemicals

Catalog number: A15609.06
5 g, Each
Thermo Scientific Chemicals

4-n-Nonylphenol, 98+%, Thermo Scientific Chemicals

Catalog number: A15609.06
5 g, Each
Quantity
Catalog number: A15609.06
also known as A15609-06
Price (USD)

Chemical Identifiers

CAS
104-40-5
IUPAC Name
4-nonylphenol
Molecular Formula
C15H24O
InChI Key
IGFHQQFPSIBGKE-UHFFFAOYSA-N
SMILES
CCCCCCCCCC1=CC=C(O)C=C1
Melting Point (clear melt)
42-48?C
Form
Crystalline powder or flakes
Identification (FTIR)
Conforms
Appearance (Color)
White to cream
Assay (GC)
≥98.0%

Description

4-n-Nonylphenol is used in manufacturing antioxidants, lubricating oil additives, laundry and dish detergents, emulsifiers, and solubilizers. Also be used to produce tris(4-nonyl-phenyl) phosphite (TNPP).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Nonylphenol is used in manufacturing antioxidants, lubricating oil additives, laundry and dish detergents, emulsifiers, and solubilizers. Also be used to produce tris(4-nonyl-phenyl) phosphite (TNPP).

Solubility
Sparingly Soluble (0.020 g/L) at 25°C.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text