4-Iodotoluene, 98%
4-Iodotoluene, 98%
4-Iodotoluene, 98%
Thermo Scientific Chemicals

4-Iodotoluene, 98%

CAS: 624-31-7 | C7H7I | 218.037 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A15631.36
also known as A15631-36
Price (EUR)
400,00
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (EUR)
400,00
Each
Add to cart
Chemical Identifiers
CAS624-31-7
IUPAC Name1-iodo-4-methylbenzene
Molecular FormulaC7H7I
InChI KeyUDHAWRUAECEBHC-UHFFFAOYSA-N
SMILESCC1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)30-39?C
Appearance (Color)White to yellow
Assay (GC)≥97.5%
FormCrystalline or fused solid
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Iodotoluene is used in wide range of medicals industrial applications as well as in human and animal nutrition products such as antiseptics and disinfectants, pharmaceutical intermediates, polarizing films for liquid crystal display [LCD] chemicals.

Solubility
Solubility in methanol is almost transparent.

Notes
Light sensitive. Store in dark. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Wancheng Zhu, et al. (Ni,Mg)3Si2O5(OH)4 solid-solution nanotubes supported by sub-0.06 wt % palladium as a robust high-efficiency catalyst for Suzuki-Miyaura cross-coupling reactions.Inorg Chem.,2012,51(11), 6020-6031.
  2. Ying-Chieh Wong, et al. Cobalt-catalyzed aryl-sulfur bond formation.Org Lett.,2006,8(24), 5613-5616.