Thermo Scientific Chemicals

3-Fluorophenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A15635.14
25 g, Each
Thermo Scientific Chemicals

3-Fluorophenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A15635.14
25 g, Each
Quantity
Catalog number: A15635.14
also known as A15635-14
Price (USD)
Price: 178.00
Online price: 152.65
Your price:
Quantity
-

Chemical Identifiers

CAS
331-25-9
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder or fused solid or flakes
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (Silylated GC)
≥97.5%
Water Content (Karl Fischer Titration)
≤1.5%

Description

3-Fluorophenylacetic acid has been used as building block to synthesize the pentaamine and bis-heterocyclic libraries. It is also used as a medicine intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Fluorophenylacetic acid has been used as building block to synthesize the pentaamine and bis-heterocyclic libraries. It is also used as a medicine intermediates.

Solubility
Slightly soluble in chloroform and methanol. Insoluble in water.

Notes
Store at room temperature. Store in cool dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text