Chlorodimethylphenylsilane, 97+%
Chlorodimethylphenylsilane, 97+%
Chlorodimethylphenylsilane, 97+%
Thermo Scientific Chemicals

Chlorodimethylphenylsilane, 97+%

CAS: 768-33-2 | C8H11ClSi | 170.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15638.06
also known as A15638-06
Price (EUR)
50,50
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
50,50
Each
Chemical Identifiers
CAS768-33-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥95.0%
Identification (FTIR)Conforms
Refractive Index1.5065-1.5105 @ 20°C
Chlorodimethylphenylsilane is used in the synthesis of enantioenriched allenylsilanes through an ortho-ester Claisen rearrangement of chiral and silylpropargylic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorodimethylphenylsilane is used in the synthesis of enantioenriched allenylsilanes through an ortho-ester Claisen rearrangement of chiral and silylpropargylic alcohols.

Solubility
Miscible with tetrahydrofuran, ether and hexane.

Notes
Incompatible with strong oxidizing agents, acids, bases, and moisture.
RUO – Research Use Only

General References:

  1. Boukachabia, M.; Vriamont, N.; Lambin, D.; Riant, O.; Aribi-Zouioueche, L. Synthesis and reactions of donor cyclopropanes: efficient routes to cis- and trans-tetrahydrofurans. Tetrahedron 2015, 71 (39), 7386-7414.
  2. Dunn, J.; Dobbs, A. P. Chlorobenzene Poisoning and Recovery of Platinum-Based Cathodes in Proton Exchange Membrane Fuel Cells. J. Phys. Chem. C 2015, 119 (35), 20328-20338.