Thermo Scientific Chemicals

Ethyl 4-chloroacetoacetate, 97%, Thermo Scientific Chemicals

Catalog number: A15641.18
50 g, Each
Thermo Scientific Chemicals

Ethyl 4-chloroacetoacetate, 97%, Thermo Scientific Chemicals

Catalog number: A15641.18
50 g, Each
Quantity
Catalog number: A15641.18
also known as A15641-18
Price (USD)
Price: 34.40
Online price: 32.65
Your price:
Quantity
-

Chemical Identifiers

CAS
638-07-3
IUPAC Name
ethyl 4-chloro-3-oxobutanoate
Molecular Formula
C6H9ClO3
InChI Key
OHLRLMWUFVDREV-UHFFFAOYSA-N
SMILES
CCOC(=O)CC(=O)CCl
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.4500-1.4550 @ 20?C

Description

Ethyl 4-chloroacetoacetate is used as a precursor in the preparation of phosphorous ylides. It finds application in regular film coating, pharmaceutical coating and formulation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-chloroacetoacetate is used as a precursor in the preparation of phosphorous ylides. It finds application in regular film coating, pharmaceutical coating and formulation.

Solubility
Miscible with water and most organic solvents.

Notes
Incompatible with strong acids, strong bases, reducing agents and strong oxidizing agents. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text