3,4-Dichlorophenylacetic acid, 98%
3,4-Dichlorophenylacetic acid, 98%
3,4-Dichlorophenylacetic acid, 98%
3,4-Dichlorophenylacetic acid, 98%
Thermo Scientific Chemicals

3,4-Dichlorophenylacetic acid, 98%

CAS: 5807-30-7 | C8H6Cl2O2 | 205.034 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15653.22
also known as A15653-22
Price (EUR)
243,00
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
243,00
Each
Add to cart
Chemical Identifiers
CAS5807-30-7
IUPAC Name2-(3,4-dichlorophenyl)acetic acid
Molecular FormulaC8H6Cl2O2
InChI KeyZOUPGSMSNQLUNW-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=C(Cl)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥97.5%
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)84.0-91.0?C
FormCrystals or powder or crystalline powder or lumps
It is used in the parallel solid phase synthesis of tetra substituted diethylenetriamines via selective amide alkylation and exhaustive reduction of N-acylated dipeptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the parallel solid phase synthesis of tetra substituted diethylenetriamines via selective amide alkylation and exhaustive reduction of N-acylated dipeptides.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Jacob Szmuszkovicz, et al. Benzeneacetamide amines: structurally novel non-m.mu. opioids.J. Med. Chem.,1982,25(10), 1125-1126.
  2. Adel Nefzi, et al. Parallel solid phase synthesis of tetrasubstituted diethylenetriamines via selective amide alkylation and exhaustive reduction of N-acylated dipeptides.Tetrahedron.,1999,55(2), 335-344.