SSEA-1 / CD15 Antibody (MEM-158), APC conjugate, Molecular Probes™

Catalog number: A15700

Invitrogen™  Related applications: Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

SSEA-1 (stage-specific embryonic antigen-1) is a cell surface antigen, expressed in murine embryonic stem cells, embryonal carcinoma cells, and induced pluripotent stem cells (iPS). SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reactivity: Human
Validated Application: Flow Cytometry
Host Species: Mouse
SubClass (Isotype): IgM
Clonality: Monoclonal
Clone ID: MEM-158
Antibody Fragment: Whole Antibody
Target Gene: fut4
Gene ID: 14345
Gene Aliases: AI451562, CD15, FAL, FucT-IV, LeX, SSEA-1, Ssea1
Label or Dye: APC (Allophycocyanin)
Excitation⁄Emission (nm): 650⁄660
Excitation Class: Visible
Form: Liquid
Number of Reactions: 100 Tests
For Use With (Equipment): Attune® Acoustic Focusing Cytometer
Product Size: 100 tests
Comments: Suggested use is 10 µl antibody /100 µl sample.
Other Additives: Sodium Azide Preservative

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products