Thermo Scientific Chemicals

Thiophene-3-carboxylic acid, 99%, Thermo Scientific Chemicals

Catalog number: A15712.22
100 g, Each
Thermo Scientific Chemicals

Thiophene-3-carboxylic acid, 99%, Thermo Scientific Chemicals

Catalog number: A15712.22
100 g, Each
Quantity
Catalog number: A15712.22
also known as A15712-22
Price (USD)
Price: 1,005.00
Online price: 877.65
Your price:
Quantity
-

Chemical Identifiers

CAS
88-13-1
IUPAC Name
thiophene-3-carboxylic acid
Molecular Formula
C5H4O2S
InChI Key
YNVOMSDITJMNET-UHFFFAOYSA-N
SMILES
OC(=O)C1=CSC=C1
Assay (HPLC)
≥98.5%
Melting Point (clear melt)
137.0-143.0?C
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

3-Thiophenecarboxylic acid has been used in the preparation of novel acetylene monomer, required for the synthesis of poly(2-methylbut-2-enyl thiophene-3-carboxylate) and oxazolines and oxazoles. It is also used as a lading compound for the development of a clinic useful D-amino acid inhibitor and have the potential to sever as active site proves to elucidate the structure-function relationships of D-amino acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Thiophenecarboxylic acid has been used in the preparation of novel acetylene monomer, required for the synthesis of poly(2-methylbut-2-enyl thiophene-3-carboxylate) and oxazolines and oxazoles. It is also used as a lading compound for the development of a clinic useful D-amino acid inhibitor and have the potential to sever as active site proves to elucidate the structure-function relationships of D-amino acids.

Solubility
Soluble in water at 20°C 4.3 g/L.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text