Hydroxymethanesulfinic acid sodium salt hydrate, tech 85%, cont. up to 5% sodium sulfite (dry wt.)
Hydroxymethanesulfinic acid sodium salt hydrate, tech 85%, cont. up to 5% sodium sulfite (dry wt.)
Hydroxymethanesulfinic acid sodium salt hydrate, tech 85%, cont. up to 5% sodium sulfite (dry wt.)
Thermo Scientific Chemicals

Hydroxymethanesulfinic acid sodium salt hydrate, tech 85%, cont. up to 5% sodium sulfite (dry wt.)

CAS: 149-44-0 | CH3NaO3S | 118.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2500 g
Catalog number A15731.0E
also known as A15731-0E
Price (USD)
130.65
Online Exclusive
145.00
Save 14.35 (10%)
Each
Quantity:
2500 g
Request bulk or custom format
Price (USD)
130.65
Online Exclusive
145.00
Save 14.35 (10%)
Each
Chemical Identifiers
CAS149-44-0
IUPAC Namesodium hydroxymethanesulfinate
Molecular FormulaCH3NaO3S
InChI KeyXWGJFPHUCFXLBL-UHFFFAOYSA-M
SMILES[Na+].OCS([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Formpowder
CommentAlkalinity as NaOH < 2.0%
Sulfated Ash> 50.0%
Appearance (Color)White
Assay (ex Sulphated Ash)> 83%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only