2-(Ethylamino)ethanol is an efficient compound used for inhibiting corrosion. This high activity of the inhibitor may be interpreted both by its ability to stabilize the hydroxide surface film and by its adsorption onto bare sites of the metal.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(Ethylamino)ethanol is an efficient compound used for inhibiting corrosion. This high activity of the inhibitor may be interpreted both by its ability to stabilize the hydroxide surface film and by its adsorption onto bare sites of the metal.
Solubility
Miscible in water.
Notes
Store at room temperature. Keep away from oxidizing agents and acids.
RUO – Research Use Only
General References:
- M. Dupra.; T.; N. Bui.; F. Dabosi. Corrosion Inhibition of a Carbon Steel by 2-Ethylamino-Ethanol in Aerated 3% NaCI Solutions - Effect of pH. Corrosion. 1979, 35 (9), 392-397 .
- Estrella Álvarez.; Diego Gómez-Díaz.; M. Dolores La Rubia.; osé M. Navaza. Densities and Viscosities of Aqueous Ternary Mixtures of 2-(Methylamino)ethanol and 2-(Ethylamino)ethanol with Diethanolamine, Triethanolamine, N-Methyldiethanolamine, or 2-Amino-1-methyl-1-propanol from 298.15 to 323.15 K.J. Chem. Eng. Data. 2006, 51 (3),955-962 .